Merck
  • Inhibition of aerobic metabolic cis-1,2-di-chloroethene biodegradation by other chloroethenes.
Inhibition of aerobic metabolic cis-1,2-di-chloroethene biodegradation by other chloroethenes.
Water research (2010-01-19)
He-Ping Zhao, Kathrin R Schmidt, Andreas Tiehm
ABSTRACT
The presence of other chloroethenes influences aerobic metabolic biodegradation of cis-1,2-dichloroethene (cDCE). A new metabolically cDCE degrading enrichment culture was identified as also being capable of degrading vinyl chloride (VC), but not 1,1-dichloroethene (1,1DCE), trans-1,2-dichloroethene (tDCE), trichloroethene (TCE), or tetrachloroethene (PCE). The fastest degradation of cDCE was observed in the absence of any other chloroethene. In the presence of a second chloroethene (40-90 microM), the rate of cDCE (60 microM) degradation decreased in the following order: cDCE (+PCE) > cDCE (+tDCE) > cDCE (+VC)>cDCE (+1,1DCE) approximately cDCE (+TCE). With increasing concentrations of VC, ranging from 10 to 110 microM, the rate of cDCE (60 microM) degradation decreased. This study demonstrates that the inhibiting effects of chloroethene mixtures have to be considered during laboratory studies and bioremediation approaches based on metabolic cDCE degradation.
Product Description
Supelco
cis-1,2-Dichloroethene, analytical standard
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Supelco
trans-1,2-Dichloroethylene, analytical standard
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol