Merck
Cucurbituril complexes cross the cell membrane.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2009-12-22)
Pedro Montes-Navajas, María González-Béjar, J C Scaiano, Hermenegildo García
ABSTRACT
Cucurbiturils (CBs) of the appropriate size (CB[7] and CB[8]) form strong guest-host complexes in phosphate buffer solution (PBS) with acridine orange (AO) and pyronine Y (PYY) with 1 : 1 and 2 : 1 stoichiometries for CB[7] and CB[8] complexes, respectively. Binding constants in the range 0.87-1.60 x 10(6) M(-1) and 5.2-6.3 x 10(13) M(-2) were determined by titration with fluorescence spectroscopy for 1 : 1 and 2 : 1 complexes, respectively. These binding constants in PBS and the eight-fold excess of CBs minimize the presence of free dye in solution and also stabilize the host-guest complex in the culture medium. Images showing that the CB complexes can cross the cell membrane of 3T3 cells have been acquired using fluorescence microscopy. Given the current importance of supramolecular CB complexes and the search for new drug delivery systems, the present findings open avenues for the use of CBs as nanocapsules to transport drugs into the cells.
Sigma-Aldrich
Pyronin Y, for microscopy (Bot., Fl., Hist.)
Sigma-Aldrich
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
Sigma-Aldrich
Pyronin Y, for NA electrophoresis