Merck
  • A method to build an analytic model of the 10B(n,alpha)7Li reaction rate space distribution for boron neutron capture therapy (BNCT).
A method to build an analytic model of the 10B(n,alpha)7Li reaction rate space distribution for boron neutron capture therapy (BNCT).
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2009-04-21)
Josselin Morand, Raymond Moss, Sabet Hachem, Wolfgang Sauerwein
ABSTRACT
This work provides the basis of a methodology to build a deterministic model for the spatial distribution of the (10)B(n,alpha)(7)Li reaction rate in boron neutron capture therapy (BNCT), as a function of space variables, boron concentration and beam incidence angle in homogeneous isotropic environments but also in different heterogeneous environments. Building the analytic function in a simple homogeneous environment with numerical methods leads to a mathematical formulation of the (10)B(n,alpha)(7)Li reactions rate.
Sigma-Aldrich
Lithium tetraborate, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium tetraborate, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium metaborate, ACS reagent, ≥98.0%
Sigma-Aldrich
Lithium metaborate, 99.9% trace metals basis
Sigma-Aldrich
Lithium metaborate, 99.995% trace metals basis