Merck
  • Toward a full characterization of native starch: separation and detection by size-exclusion chromatography.
Toward a full characterization of native starch: separation and detection by size-exclusion chromatography.
Journal of chromatography. A (2008-08-30)
Ngoc-Ly Hoang, Antonin Landolfi, Anastasiya Kravchuk, Etienne Girard, Jonathan Peate, Javier M Hernandez, Marianne Gaborieau, Olena Kravchuk, Robert G Gilbert, Yohann Guillaneuf, Patrice Castignolles
ABSTRACT
The structure of starch molecules is relevant to nutrition and industrial applications. Size-exclusion chromatography (SEC, also known as GPC) of native starch generally suffers non-satisfactory repeatability and reproducibility of the dissolution and separation. This work combines two polar organic solvents: dimethylsulfoxide for complete dissolution and dimethylacetamide to limit shear degradation. The separation is as repeatable as that of polystyrene standards performing dissolution and separation at 80 degrees C. Successful covalent-labeling on the glucose unit is claimed to be published here for the first time in non-degradative conditions and allows the use of UV detector with significantly higher sensitivity than with a refractometer.
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O