Merck
  • The mechanism of ethylene glycol ether reproductive and developmental toxicity and evidence for adverse effects in humans.
The mechanism of ethylene glycol ether reproductive and developmental toxicity and evidence for adverse effects in humans.
Toxicology letters (2005-02-12)
Frank Welsch
ABSTRACT
Numerous experimental studies have established that only a few among the large family of ethylene glycol ethers (EGEs) elicit toxicity on reproduction in either gender. Notable are the monomethyl (EGME) and monoethyl (EGEE) ethers and their respective acetate esters whose production volumes have dramatically declined. Oxidation to the respective monoalkoxy acids is a prerequisite for toxicity. The most potent EGE reproductive toxicant is EGME (via 2-methoxyacetic acid; MAA), which elicits developmental phase-specific insults on either conceptus or on testes. Toxicity at either target site is markedly attenuated by simple physiological compounds such as acetate, formate, glycine, D-glucose and serine. Lack of solid EGME occupational exposure data and the need to improve the scientific foundations for animal data extrapolations, prompted the development of physiologically based pharmacokinetic (PBPK) models for pregnancy application. Interspecies (mouse-rat) and different exposure routes (including inhalation) were experimentally validated. Such PBPK models were then extrapolated to potential occupational exposures, using rather limited human MAA pharmacokinetic data. PBPK model predictions of human blood levels upon simulated inhalation exposure to the 5 ppm threshold limit value (TLV) for 8 h were approximately 60 microM were well below those causing adverse effects in pregnant mice or rats. This conclusion concurs with the lack of objective analytical chemistry data for EGME/MAA in occupational settings, regardless of the potential route of exposure. There are no exposure data that can be linked in a cause-and-effect association to adverse human reproductive outcomes.
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Sigma-Aldrich
2-Methoxyethanol, contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%
Sigma-Aldrich
2-Methoxyethanol, ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer
Supelco
2-Methoxyethanol, analytical standard
Sigma-Aldrich
2-Methoxyethanol, suitable for HPLC, ≥99.9%