Merck
  • Parallel synthesis of oxazolines and thiazolines by tandem condensation-cyclodehydration of carboxylic acids with amino alcohols and aminothiols.
Parallel synthesis of oxazolines and thiazolines by tandem condensation-cyclodehydration of carboxylic acids with amino alcohols and aminothiols.
Journal of combinatorial chemistry (2002-11-12)
Peter Wipf, Xiaodong Wang
ABSTRACT
A combinatorial library of oxazolines and thiazolines was synthesized in moderate to excellent yields using a newly developed methodology. Free carboxylic acids were directly condensed with amino alcohols and aminothiols in the presence of 3-nitrophenylboronic acid as a dehydration catalyst. The library synthesis illustrates the scope of this process: After traditional reaction optimization, a total of 17 oxazolines and 6 thiazolines were successfully prepared in a 24-reaction setup in a Radleys GreenHouse parallel synthesizer. The yields of the parallel reactions ranged from moderate to excellent, depending largely on carboxylate reactivity and functionalization, and generally exceeded those of the traditional reaction setup. The target compounds were isolated in high purities (average purity is 96% according to GC analysis) after passing reaction mixtures through short PrepSep SPE Florisil cartridges.
Sigma-Aldrich
3-Nitrophenylboronic acid, ≥97%