Merck
  • Dual-emissive quantum dots for multispectral intraoperative fluorescence imaging.
Dual-emissive quantum dots for multispectral intraoperative fluorescence imaging.
Biomaterials (2010-07-14)
Patrick T K Chin, Tessa Buckle, Arantxa Aguirre de Miguel, Stefan C J Meskers, René A J Janssen, Fijs W B van Leeuwen
ABSTRACT
Fluorescence molecular imaging is rapidly increasing its popularity in image guided surgery applications. To help develop its full surgical potential it remains a challenge to generate dual-emissive imaging agents that allow for combined visible assessment and sensitive camera based imaging. To this end, we now describe multispectral InP/ZnS quantum dots (QDs) that exhibit a bright visible green/yellow exciton emission combined with a long-lived far red defect emission. The intensity of the latter emission was enhanced by X-ray irradiation and allows for: 1) inverted QD density dependent defect emission intensity, showing improved efficacies at lower QD densities, and 2) detection without direct illumination and interference from autofluorescence.
Sigma-Aldrich
InP/ZnS quantum dots, stabilized with oleylamine ligands, fluorescence λem 530 nm, 5 mg/mL in toluene
Sigma-Aldrich
InP/ZnS quantum dots, stabilized with oleylamine ligands, fluorescence λem 590 nm, 5 mg/mL in toluene
Sigma-Aldrich
InP/ZnS quantum dots, stabilized with oleylamine ligands, fluorescence λem 620 nm, 5 mg/mL in toluene
Sigma-Aldrich
InP/ZnS quantum dots, stabilized with oleylamine ligands, fluorescence λem 650 nm, 5 mg/mL in toluene
Sigma-Aldrich
InP/ZnS quantum dots, stabilized with oleylamine ligands, fluorescence λem 560 nm, 5 mg/mL in toluene