Merck
  • MicroRNA-93 promotes the pathogenesis of glaucoma by inhibiting matrix metalloproteinases as well as up-regulating extracellular matrix and Rho/ROCK signaling pathways.
MicroRNA-93 promotes the pathogenesis of glaucoma by inhibiting matrix metalloproteinases as well as up-regulating extracellular matrix and Rho/ROCK signaling pathways.
Heliyon (2023-12-04)
Manhua Xu, Yanxi Wang, Juan Zhou, Xun Zhang, Yinggui Yu, Kaiming Li
ABSTRACT
To investigate the effect and potential molecular mechanism of microRNA-93 (miR-93) on retinal ganglion cells (RGCs) apoptosis as well as retinal damage in acute glaucoma mice. RGCs apoptosis were induced by oxygen-glucose deprivation and reperfusion (OGD/R). The pro-apoptotic effect of miR-93 was evaluated by transfecting miR-93 mimics or miR-93 inhibitor into OGD/R-induced RGCs. The viability and apoptosis of RGCs were determined by MTT assay and flow cytometry. Mouse model of acute glaucoma were successfully induced via high intraocular pressure (IOP), and then these model animals were randomly divided into vehicle group, miR-93 mimics group or miR-93 inhibitor group (n = 10), using healthy mice as normal control. Histopathologic changes of retinal tissue were evaluated by Hematoxylin and Eosin (H&E) staining method. Moreover, cell counts of retinal ganglion cell layer and mean thickness of different layers were also determined. Quantitative real-time PCR (qPCR) and western blotting analysis were used to detect the mRNA and protein expression levels of extracellular matrix (ECM), matrix metalloproteinases (MMPs) and Rho/ROCK signaling pathway. miR-93 mimics significantly decreased or promoted the viability and apoptosis of OGD/R-induced RGCs, respectively. In addition, miR-93 mimics significantly exacerbated the degree of retinal tissue damage in mice with acute glaucoma, which was accompanied by a decrease in the number of ganglion cell layer (GCL) cells and the thickness of different tissue layers. Moreover, miR-93 mimics significantly increased IOP in mice with acute glaucoma. Significantly, miR-93 inhibitors significantly reversed the above changes. In addition, results of Western blot analysis showed that miR-93 mimics increased and decreased the expression of ECM-associated and MMP-associated proteins, respectively, by activating the Rho/ROCK signaling pathway. In contrast, miR-93 significantly decreased and increased the expression of ECM-associated and MMP-associated proteins, and suppressed the expression of Rho/ROCK signaling pathway-related proteins. miR-93 can promote the development of glaucoma by activating Rho/ROCK signaling pathway to mediate the accumulation of ECM-related proteins as well as the down-regulation of MMP-related proteins.
Sigma-Aldrich
Anti-RHOA antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-ROCK2 (pTyr722) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TGF β1 antibody produced in rabbit, affinity isolated antibody