Merck
  • The transcription factor TCFL5 responds to A-MYB to elaborate the male meiotic program in mice.
The transcription factor TCFL5 responds to A-MYB to elaborate the male meiotic program in mice.
Reproduction (Cambridge, England) (2022-11-18)
Katharine Cecchini, Adriano Biasini, Tianxiong Yu, Martin Säflund, Haiwei Mou, Amena Arif, Atiyeh Eghbali, Cansu Colpan, Ildar Gainetdinov, Dirk G de Rooij, Zhiping Weng, Phillip D Zamore, Deniz M Özata
ABSTRACT
The testis-specific transcription factor, TCFL5, expressed in pachytene spermatocytes regulates the meiotic gene expression program in collaboration with the transcription factor A-MYB. In male mice, the transcription factors STRA8 and MEISON initiate meiosis I. We report that STRA8/MEISON activates the transcription factors A-MYB and TCFL5, which together reprogram gene expression after spermatogonia enter into meiosis. TCFL5 promotes the transcription of genes required for meiosis, mRNA turnover, miR-34/449 production, meiotic exit, and spermiogenesis. This transcriptional architecture is conserved in rhesus macaque, suggesting TCFL5 plays a central role in meiosis and spermiogenesis in placental mammals. Tcfl5em1/em1 mutants are sterile, and spermatogenesis arrests at the mid- or late-pachytene stage of meiosis. Moreover, Tcfl5+/em1 mutants produce fewer motile sperm.
Sigma-Aldrich
Anti-MYBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-BrdU Antibody, clone PRB-1, biotin conjugated, clone PRB-1, Chemicon®, from mouse
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Anti-TCFL5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution