Merck
  • Inherited ARPC5 mutations cause an actinopathy impairing cell motility and disrupting cytokine signaling.
Inherited ARPC5 mutations cause an actinopathy impairing cell motility and disrupting cytokine signaling.
Nature communications (2023-06-23)
Cristiane J Nunes-Santos, HyeSun Kuehn, Brigette Boast, SuJin Hwang, Douglas B Kuhns, Jennifer Stoddard, Julie E Niemela, Danielle L Fink, Stefania Pittaluga, Mones Abu-Asab, John S Davies, Valarie A Barr, Tomoki Kawai, Ottavia M Delmonte, Marita Bosticardo, Mary Garofalo, Magda Carneiro-Sampaio, Raz Somech, Mohammad Gharagozlou, Nima Parvaneh, Lawrence E Samelson, Thomas A Fleisher, Anne Puel, Luigi D Notarangelo, Bertrand Boisson, Jean-Laurent Casanova, Beata Derfalvi, Sergio D Rosenzweig
ABSTRACT
We describe the first cases of germline biallelic null mutations in ARPC5, part of the Arp2/3 actin nucleator complex, in two unrelated patients presenting with recurrent and severe infections, early-onset autoimmunity, inflammation, and dysmorphisms. This defect compromises multiple cell lineages and functions, and when protein expression is reestablished in-vitro, the Arp2/3 complex conformation and functions are rescued. As part of the pathophysiological evaluation, we also show that interleukin (IL)-6 signaling is distinctively impacted in this syndrome. Disruption of IL-6 classical but not trans-signaling highlights their differential roles in the disease and offers perspectives for therapeutic molecular targets.
Sigma-Aldrich
Anti-ARPC1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARPC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARPC1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ARPC4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ARPC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution