Merck
  • Structure elucidation and biological activities of perylenequinones from an Alternaria species.
Structure elucidation and biological activities of perylenequinones from an Alternaria species.
Mycotoxin research (2023-06-23)
Anna Kiefer, Marcel Arnholdt, Viktoria Grimm, Leander Geske, Jonathan Groß, Nina Vierengel, Till Opatz, Gerhard Erkel
ABSTRACT
The KEAP1-Nrf2/ARE pathway is a pivotal cytoprotective regulator against oxidative stress which plays an important role in the development of many inflammatory diseases and cancer. Activation of the Nrf2 transcription factor by oxidative stress or electrophiles regulates antioxidant response element (ARE)-dependent transcription of antioxidative, detoxifying, and anti-inflammatory proteins. Therefore, modulators of the KEAP1-Nrf2/ARE pathway have received considerable interest as therapeutics to protect against diseases where oxidative stress constitutes the underlying pathophysiology. In a search for fungal secondary metabolites affecting the Nrf2/ARE-dependent expression of a luciferase reporter gene in BEAS-2B cells, three new perylenequinones, compounds 1, 2, and 3, together with altertoxin-I (ATX-I), were isolated from fermentations of an Alternaria species. The structures of the compounds were elucidated by a combination of one- and two-dimensional NMR spectroscopy and mass spectrometry. Compound 1 and ATX-I exhibited strong cytotoxic effects with LC50-values of 3.8 µM and 6.43 µM, respectively, whereas compound 3 showed no cytotoxic effects up to 100 µM on BEAS-2B cells. ATX-I induced ARE-dependent luciferase expression approximately fivefold and compound 1 approximately 2.6-fold at a concentration of 3 µM in transiently transfected BEAS-2B cells. In addition, compound 1 and ATX-I exhibited strong oxidative effects, whereas compound 3 did not show significant oxidative properties. For compound 1 and ATX-I, a strong upregulation of heme oxygenase-1 could be observed on mRNA and protein level in treated BEAS-2B cells. Moreover, compound 3 significantly decreased sod3 mRNA levels after induction of oxidative stress with benzoquinone.
Sigma-Aldrich
Anti-NRF2 Antibody, clone 6H11.1, clone 6H11.1, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution