Merck
  • Post-acute sequelae of COVID-19 is characterized by diminished peripheral CD8+β7 integrin+ T cells and anti-SARS-CoV-2 IgA response.
Post-acute sequelae of COVID-19 is characterized by diminished peripheral CD8+β7 integrin+ T cells and anti-SARS-CoV-2 IgA response.
Nature communications (2023-03-31)
André Santa Cruz, Ana Mendes-Frias, Marne Azarias-da-Silva, Sónia André, Ana Isabel Oliveira, Olga Pires, Marta Mendes, Bárbara Oliveira, Marta Braga, Joana Rita Lopes, Rui Domingues, Ricardo Costa, Luís Neves Silva, Ana Rita Matos, Cristina Ângela, Patrício Costa, Alexandre Carvalho, Carlos Capela, Jorge Pedrosa, António Gil Castro, Jérôme Estaquier, Ricardo Silvestre
ABSTRACT
Several millions of individuals are estimated to develop post-acute sequelae SARS-CoV-2 condition (PASC) that persists for months after infection. Here we evaluate the immune response in convalescent individuals with PASC compared to convalescent asymptomatic and uninfected participants, six months following their COVID-19 diagnosis. Both convalescent asymptomatic and PASC cases are characterised by higher CD8+ T cell percentages, however, the proportion of blood CD8+ T cells expressing the mucosal homing receptor β7 is low in PASC patients. CD8 T cells show increased expression of PD-1, perforin and granzyme B in PASC, and the plasma levels of type I and type III (mucosal) interferons are elevated. The humoral response is characterized by higher levels of IgA against the N and S viral proteins, particularly in those individuals who had severe acute disease. Our results also show that consistently elevated levels of IL-6, IL-8/CXCL8 and IP-10/CXCL10 during acute disease increase the risk to develop PASC. In summary, our study indicates that PASC is defined by persisting immunological dysfunction as late as six months following SARS-CoV-2 infection, including alterations in mucosal immune parameters, redistribution of mucosal CD8+β7Integrin+ T cells and IgA, indicative of potential viral persistence and mucosal involvement in the etiopathology of PASC.
Sigma-Aldrich
Anti-Human IgA (α-chain specific), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Goat Anti-Human IgM, μ-Chain Specific Peroxidase Conjugate, liquid, Calbiochem®