Merck
  • Cystine Induced-mTORC2 Activation through Promoting Sin1 Phosphorylation to Suppress Cancer Cell Ferroptosis.
Cystine Induced-mTORC2 Activation through Promoting Sin1 Phosphorylation to Suppress Cancer Cell Ferroptosis.
Molecular nutrition & food research (2022-10-04)
GuoYan Wang, Lei Chen, SenLin Qin, YiNing Zheng, Chao Xia, JunHu Yao, Ping Wang, Lu Deng
ABSTRACT
Mechanistic target of rapamycin (mTOR) serves as a central signaling node in the coordination of cell growth and metabolism, and it functions via two distinct complexes, namely, mTOR complex 1 (mTORC1) and mTORC2. mTORC1 plays a crucial role in sensing amino acids, whereas mTORC2 involves in sensing growth factors. However, it remains largely unclear whether mTORC2 can sense amino acids and the mechanism by which amino acids regulate mTORC2 has not been studied. After treating cells with indicated concentration of amino acids for different time, it is found that the mTORC2 activation is significantly increased in response to amino acids stimulation, especially cystine. Particularly, knockdown solute carrier family 7 member 11 (SLC7A11) by siRNA shows that SLC7A11-mediated cystine uptake is responsible for activating mTORC2. Mechanistically, the study finds that p38 is activated in response to cystine stimulation, and co-immunoprecipitation (Co-IP) experiments suggest that p38 regulates the assembly of components within mTORC2 by mediating the phosphorylation of the mTORC2 subunit mitogen-activated protein kinase-interacting protein 1 (Sin1) in a cystine-dependent manner. Finally, combined with inducers and inhibitors of ferroptosis and cell viability assay, the study observes that cystine-mediated regulation of the p38-Sin1-mTOR-AKT pathway induces resistance to ferroptosis. These results indicate that cystine-induced activation of the p38-Sin1-mTORC2-AKT pathway suppresses ferroptosis.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
PDGF-BB Protein, Human Recombinant Animal Free, The Platelet derived growth factor (PDGF) was discovered as a major mitogenic factor present in serum but absent from plasma. Recombinant animal free human PDGF-BB is manufactured using all non-animal reagents.
Sigma-Aldrich
mTOR Inhibitor XI, Torin1, mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)