Merck
  • An androgen receptor switch underlies lineage infidelity in treatment-resistant prostate cancer.
An androgen receptor switch underlies lineage infidelity in treatment-resistant prostate cancer.
Nature cell biology (2021-09-08)
Alastair Davies, Shaghayegh Nouruzi, Dwaipayan Ganguli, Takeshi Namekawa, Daksh Thaper, Simon Linder, Fatih Karaoğlanoğlu, Meltem E Omur, Soojin Kim, Maxim Kobelev, Sahil Kumar, Olena Sivak, Chiara Bostock, Jennifer Bishop, Marlous Hoogstraat, Amina Talal, Suzan Stelloo, Henk van der Poel, Andries M Bergman, Musaddeque Ahmed, Ladan Fazli, Haojie Huang, Wayne Tilley, David Goodrich, Felix Y Feng, Martin Gleave, Housheng Hansen He, Faraz Hach, Wilbert Zwart, Himisha Beltran, Luke Selth, Amina Zoubeidi
ABSTRACT
Cancers adapt to increasingly potent targeted therapies by reprogramming their phenotype. Here we investigated such a phenomenon in prostate cancer, in which tumours can escape epithelial lineage confinement and transition to a high-plasticity state as an adaptive response to potent androgen receptor (AR) antagonism. We found that AR activity can be maintained as tumours adopt alternative lineage identities, with changes in chromatin architecture guiding AR transcriptional rerouting. The epigenetic regulator enhancer of zeste homologue 2 (EZH2) co-occupies the reprogrammed AR cistrome to transcriptionally modulate stem cell and neuronal gene networks-granting privileges associated with both fates. This function of EZH2 was associated with T350 phosphorylation and establishment of a non-canonical polycomb subcomplex. Our study provides mechanistic insights into the plasticity of the lineage-infidelity state governed by AR reprogramming that enabled us to redirect cell fate by modulating EZH2 and AR, highlighting the clinical potential of reversing resistance phenotypes.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
GSK343, ≥98% (HPLC)
Sigma-Aldrich
ChIPAb+ EED - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
A-395 hydrochloride, ≥98% (HPLC)