Merck
  • Phosphosites of the yeast centrosome component Spc110 contribute to cell cycle progression and mitotic exit.
Phosphosites of the yeast centrosome component Spc110 contribute to cell cycle progression and mitotic exit.
Biology open (2022-10-20)
Marjan Abbasi, Alexander Julner, Yan Ting Lim, Tianyun Zhao, Radoslaw Mikolaj Sobota, Victoria Menéndez-Benito
ABSTRACT
Spc110 is an essential component of the spindle pole body (SPB), the yeast equivalent of the centrosome, that recruits the γ-tubulin complex to the nuclear side of the SPB to produce the microtubules that form the mitotic spindle. Here, we identified phosphosites S11 and S36 in maternally originated Spc110 and explored their functions in vivo. Yeast expressing non-phosphorylatable Spc110S11A had a distinct spindle phenotype characterised by higher levels of α-tubulin, which was frequently asymmetrically distributed between the two SPBs. Furthermore, expression of the double mutant Spc110S11AS36A had a delayed cell cycle progression. Specifically, the final steps of mitosis were delayed in Spc110S11AS36A cells, including expression and degradation of the mitotic cyclin Clb2, disassembling the mitotic spindle and re-localizing Cdc14 to the nucleoli, resulting in late mitotic exit and entry in G1. Thus, we propose that Spc110 phosphorylation at S11 and S36 is required to regulate timely cell cycle progression in budding yeast. This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
RNase, from bovine pancreas