Merck
  • O,O'-Diester methylenediphosphonotetrathioate: synthesis, characterization, and potential applications.
O,O'-Diester methylenediphosphonotetrathioate: synthesis, characterization, and potential applications.
The Journal of organic chemistry (2012-12-05)
Aviran Amir, Alon H Sayer, Rostislav Zagalsky, Linda J W Shimon, Bilha Fischer
ABSTRACT
A new transformation of methylene-bis(phosphonic dichloride) into tetrathiobisphosphonate derivatives is reported. The reaction of methylene-bis(phosphonic dichloride) with 1,2-ethanedithiol in bromoform in the presence of AlCl(3) formed methylene-bis(1,3,2-dithiaphospholane-2-sulfide), which gave rise to O,O'-diester-methylenediphosphonotetrathioate analogues 1a-k upon reaction with phenols and alkyl alcohols in the presence of DBU. Reaction mechanisms are proposed, and all products were characterized by (31)P, (13)C, and (1)H NMR. An X-ray crystal structure was obtained for intermediate 2. The potential of the novel scaffold for selective coordination of metal-ions was examined by coordination of Hg(II) and Pb(II) by 1f, as determined by FT-IR, and chelation of Zn(II), but not Ca(II), by 1b, as determined by (31)P/(1)H NMR. UV-vis measurements of 1g-Ni(II) mixture revealed a 2:1 ligand:metal complex. These derivatives are potential antioxidants, and their ability to inhibit ·OH formation in Fenton reactions was quantified by ESR measurements. Analogue 1g proved to be a most potent antioxidant (IC(50) 53 μM), inhibiting the Cu(I)-catalyzed Fenton reaction at lower concentrations than GSH, ascorbic acid, and EDTA. Analogue 1c inhibited the Fe(II)-catalyzed Fenton reaction at about the same concentrations as ascorbic acid (IC(50) 83 vs 93 μM). In summary, the novel compounds, 1a-k, proved to chelate various borderline/soft Lewis acid metal-ions, and may be useful as antioxidants and metal extractors.
Sigma-Aldrich
Methylenebis(phosphonic dichloride), 97%
Pricing and availability is not currently available.