Merck
  • Gain of gene regulatory network interconnectivity at the origin of vertebrates.
Gain of gene regulatory network interconnectivity at the origin of vertebrates.
Proceedings of the National Academy of Sciences of the United States of America (2022-03-10)
Alejandro Gil-Gálvez, Sandra Jiménez-Gancedo, Alberto Pérez-Posada, Martin Franke, Rafael D Acemel, Che-Yi Lin, Cindy Chou, Yi-Hsien Su, Jr-Kai Yu, Stephanie Bertrand, Michael Schubert, Héctor Escrivá, Juan J Tena, José Luis Gómez-Skarmeta
ABSTRACT
SignificanceIn this manuscript, we address an essential question in developmental and evolutionary biology: How have changes in gene regulatory networks contributed to the invertebrate-to-vertebrate transition? To address this issue, we perturbed four signaling pathways critical for body plan formation in the cephalochordate amphioxus and in zebrafish and compared the effects of such perturbations on gene expression and gene regulation in both species. Our data reveal that many developmental genes have gained response to these signaling pathways in the vertebrate lineage. Moreover, we show that the interconnectivity between these pathways is much higher in zebrafish than in amphioxus. We conclude that this increased signaling pathway complexity likely contributed to vertebrate morphological novelties during evolution.
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
SU5402, SU5402, CAS 215543-92-3, is a cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of FGFR1 (IC₅₀ = 10-20 µM in the presence of 1 mM ATP).
Sigma-Aldrich
Pluripotin, ≥98% (HPLC)