Merck
ETV7 limits antiviral gene expression and control of influenza viruses.
Science signaling (2021-07-15)
Heather M Froggatt, Alfred T Harding, Ryan R Chaparian, Nicholas S Heaton
ABSTRACT
The type I interferon (IFN) response is an important component of the innate immune response to viral infection. Precise control of IFN responses is critical because insufficient expression of IFN-stimulated genes (ISGs) can lead to a failure to restrict viral spread, whereas excessive ISG activation can result in IFN-related pathologies. Although both positive and negative regulatory factors control the magnitude and duration of IFN signaling, it is also appreciated that several ISGs regulate aspects of the IFN response themselves. In this study, we performed a CRISPR activation screen to identify previously unknown regulators of the type I IFN response. We identified the strongly induced ISG encoding ETS variant transcription factor 7 (ETV7) as a negative regulator of the type I IFN response. However, ETV7 did not uniformly suppress ISG transcription. Instead, ETV7 preferentially targeted a subset of antiviral ISGs that were particularly important for IFN-mediated control of influenza viruses. Together, our data assign a function for ETV7 as an IFN response regulator and also identify ETV7 as a potential therapeutic target to increase innate antiviral responses and enhance IFN-based antiviral therapies.
Sigma-Aldrich
Anti-ETV7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Interferon-α A Protein, Recombinant human, Purified Recombinant Human Interferon Alpha A (Hu-IFN-alphaA & Hu-IFN-alpha2a).