Merck
  • Fas regulates the apoptosis and migration of trophoblast cells by targeting NF-κB.
Fas regulates the apoptosis and migration of trophoblast cells by targeting NF-κB.
Experimental and therapeutic medicine (2021-08-27)
Ruihong Lan, Yang Yang, Jie Song, Ling Wang, Humin Gong
ABSTRACT
Placental trophoblast apoptosis is a major pathological feature of preeclampsia. Fas has been reported to be highly expressed in the placentas of patients with preeclampsia. However, the role and underlying mechanisms of Fas in the pathogenesis of preeclampsia have not been elucidated. In the present study, the expression of Fas in JAR human choriocarcinoma cells was overexpressed and knocked down to determine the function and possible mechanism of Fas in trophoblast cells in the progression of preeclampsia. The results of flow cytometry, Cell Counting Kit-8 and Transwell assays indicated that the overexpression of Fas promoted apoptosis, suppressed viability and impaired the migration of the human trophoblast cells. In addition, western blotting revealed that the overexpression of Fas increased the expression of nuclear factor kB (NF-kB), Bax, tumor necrosis factor α (TNF-α) and interleukin-2 (IL-2), and decreased the expression of Bcl-2 at the protein level in trophoblast cells. By contrast, the knockdown of Fas decreased the apoptosis of trophoblast cells and increased their viability and migration. In addition, the knockdown of Fas suppressed the expression of NF-κB, Bax, TNF-α and IL-2, and increased the expression of Bcl-2. Notably, the overexpression of NF-κB p65 attenuated the Fas knockdown-induced inhibition of apoptosis and acceleration of migration of the trophoblast cells. The overexpression of NF-κB in trophoblast cells also reversed the reduction in Bax expression and increase in Bcl-2 expression induced by Fas knockdown in trophoblast cells. These results indicate that Fas regulates the apoptosis and migration of trophoblast cells by targeting NF-κB, which suggests that the silencing of Fas is a promising therapeutic strategy for preeclampsia.
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-2 ELISA Kit, for serum, plasma, cell culture supernatant and urine