Merck
  • Kainate receptors regulate the functional properties of young adult-born dentate granule cells.
Kainate receptors regulate the functional properties of young adult-born dentate granule cells.
Cell reports (2021-09-23)
Yiwen Zhu, John N Armstrong, Anis Contractor
ABSTRACT
Both inhibitory and excitatory neurotransmitter receptors can influence maturation and survival of adult-born neurons in the dentate gyrus; nevertheless, how these two neurotransmitter systems affect integration of new neurons into the existing circuitry is still not fully characterized. Here, we demonstrate that glutamate receptors of the kainate receptor (KAR) subfamily are expressed in adult-born dentate granule cells (abDGCs) and that, through their interaction with GABAergic signaling mechanisms, they alter the functional properties of adult-born cells during a critical period of their development. Both the intrinsic properties and synaptic connectivity of young abDGCs were affected. Timed KAR loss in a cohort of young adult-born neurons in mice disrupted their performance in a spatial discrimination task but not in a hippocampal-dependent fear conditioning task. Together, these results demonstrate the importance of KARs in the proper functional development of young abDGCs.
Sigma-Aldrich
Donkey Anti-Chicken IgY Antibody, biotin conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Gramicidin from Bacillus aneurinolyticus (Bacillus brevis), Linear polypeptide antibiotic complex. A mixture of gramicidins A, B, C, and D.
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Millipore
FluorSave Reagent