Merck
RNA Stability Measurements Using RT-qPCR in Arabidopsis Seedlings.
Bio-protocol (2021-03-05)
Tianran Jia, Brandon H Le
ABSTRACT
Steady-state mRNA levels are determined by both the rates of transcription and degradation. Regulation of mRNA stability and/or degradation are key factors that can significantly affect mRNA levels and its biological functions. mRNA stability can be measured indirectly after transcription inhibition. This protocol described a rapid and sensitive method of mRNA stability measurement through quantitative reverse transcription PCR (RT-qPCR) after inhibition of RNA transcription by cordycepin in Arabidopsis seedlings.
Sigma-Aldrich
Cordycepin, from Cordyceps militaris
Sigma-Aldrich
Murashige and Skoog Basal Medium, powder, suitable for plant cell culture
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid