Merck
Pharmacology of the GABAB receptor in amphibian retina.
Brain research (1994-10-17)
N Tian, M M Slaughter
ABSTRACT
Amacrine and ganglion cells in the amphibian retina contain GABAB, as well as GABAA, receptors. Baclofen, a GABAB agonist, hyperpolarizes the dark membrane potential of these third order neurons and makes their light responses more transient. GABAB receptors in the retina have a similar agonist profile to GABAB receptors described at other sites in the brain. Namely, preferential activation by the R-enantiomer of baclofen, and agonist sensitivity in the order 3-aminopropylphosphinic acid > baclofen > 3-aminopropylphosphonic acid. The GABAB receptor was not activated by 4-aminobutylphosphonic acid. Several antagonists, such as phaclofen, saclofen, and 2-hydroxysaclofen, were ineffective in the amphibian retina. However, CGP35348 blocked the action of applied baclofen and produced effects on the light response that were opposite to those of baclofen. Applied agonists and antagonists support the hypothesis that GABAB receptors serve to regulate the balance of sustained and transient signals to the inner retina.
Sigma-Aldrich
4-Aminobutylphosphonic acid, ≥99%
Sigma-Aldrich
3-Aminopropylphosphonic acid, 98%