Merck
  • Complete Genome Sequence of Acinetobacter calcoaceticus CA16, a Bacterium Capable of Degrading Diesel and Lignin.
Complete Genome Sequence of Acinetobacter calcoaceticus CA16, a Bacterium Capable of Degrading Diesel and Lignin.
Genome announcements (2017-06-18)
Margaret T Ho, Brian Weselowski, Ze-Chun Yuan
ABSTRACT
We report here the complete assembled genome sequence of Acinetobacter calcoaceticus CA16, which is capable of utilizing diesel and lignin as a sole carbon source. CA16 contains a 4,110,074-bp chromosome and a 5,920-bp plasmid. The assembled sequences will help elucidate potential metabolic pathways and mechanisms responsible for CA16's hydrocarbon degradation ability.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 350 purifications
Pricing and availability is not currently available.