Merck
BOB.1 controls memory B-cell fate in the germinal center reaction.
Journal of autoimmunity (2019-05-06)
Maartje J Levels, Cynthia M Fehres, Lisa G M van Baarsen, Nathalie O P van Uden, Kristine Germar, Tom G O'Toole, Iris C J Blijdorp, Johanna F Semmelink, Marieke E Doorenspleet, Arjen Q Bakker, Mikhail Krasavin, Alexey Tomilin, Sophie Brouard, Hergen Spits, Dominique L P Baeten, Nataliya G Yeremenko
ABSTRACT
During T cell-dependent (TD) germinal center (GC) responses, naïve B cells are instructed to differentiate towards GC B cells (GCBC), high-affinity long-lived plasma cells (LLPC) or memory B cells (Bmem). Alterations in the B cell-fate choice could contribute to immune dysregulation leading to the loss of self-tolerance and the initiation of autoimmune disease. Here we show that mRNA levels of the transcription regulator BOB.1 are increased in the lymph node compartment of patients with rheumatoid arthritis (RA), a prototypical autoimmune disease caused by the loss of immunological tolerance. Investigating to what extent levels of BOB.1 impact B cells during TD immune responses we found that BOB.1 has a crucial role in determining the B cell-fate decision. High BOB.1 levels promote the generation of cells with phenotypic and functional characteristics of Bmem. Mechanistically, overexpression of BOB.1 drives ABF1 and suppresses BCL6, favouring Bmem over LLPC or recycling GCBC. Low levels of BOB.1 are sufficient for LLPC but not for Bmem differentiation. Our findings demonstrate a novel role for BOB.1 in B cells during TD GC responses and suggest that its dysregulation may contribute to the pathogenesis of RA by disturbing the B cell-fate determination.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Staphylococcal enterotoxin B from Staphylococcus aureus
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Millipore
Millex-HP Filter Unit (Sterile), pore size 0.45 μm, diam. 33 mm, polyethersulfone membrane, hydrophilic, medical device