Merck
CASP9 germline mutation in a family with multiple brain tumors.
Brain pathology (Zurich, Switzerland) (2016-12-10)
Michael W Ronellenfitsch, Ji-Eun Oh, Kaishi Satomi, Koichiro Sumi, Patrick N Harter, Joachim P Steinbach, Jörg Felsberg, David Capper, Catherine Voegele, Geoffroy Durand, James McKay, Florence Le Calvez-Kelm, Jens Schittenhelm, Barbara Klink, Michel Mittelbronn, Hiroko Ohgaki
ABSTRACT
We report a novel CASP9 germline mutation that may increase susceptibility to the development of brain tumors. We identified this mutation in a family in which three brain tumors had developed within three generations, including two anaplastic astrocytomas occurring in cousins. The cousins were diagnosed at similar ages (29 and 31 years), and their tumors showed similar histological features. Genetic analysis revealed somatic IDH1 and TP53 mutations in both tumors. However, no germline TP53 mutations were detected, despite the fact that this family fulfills the criteria of Li-Fraumeni-like syndrome. Whole exome sequencing revealed a germline stop-gain mutation (R65X) in the CASP9 gene, which encodes caspase-9, a key molecule for the p53-dependent mitochondrial death pathway. This mutation was also detected in DNA extracted from blood samples from the two siblings who were each a parent of one of the affected cousins. Caspase-9 immunohistochemistry showed the absence of caspase-9 immunoreactivity in the anaplastic astrocytomas and normal brain tissues of the cousins. These observations suggest that CASP9 germline mutations may have played a role at least in part to the susceptibility of development of gliomas in this Li-Fraumeni-like family lacking a TP53 germline mutation.
Sigma-Aldrich
Anti-ATRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-CASP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution