Merck
  • Diclofenac impairs autophagic flux via oxidative stress and lysosomal dysfunction: Implications for hepatotoxicity.
Diclofenac impairs autophagic flux via oxidative stress and lysosomal dysfunction: Implications for hepatotoxicity.
Redox biology (2020-10-21)
Seung-Hwan Jung, Wonseok Lee, Seung-Hyun Park, Kang-Yo Lee, You-Jin Choi, Soohee Choi, Dongmin Kang, Sinri Kim, Tong-Shin Chang, Soon-Sun Hong, Byung-Hoon Lee
ABSTRACT
Treatment with nonsteroidal anti-inflammatory drugs (NSAIDs) is associated with various side effects, including cardiovascular and hepatic disorders. Studies suggest that mitochondrial damage and oxidative stress are important mediators of toxicity, yet the underlying mechanisms are poorly understood. In this study, we identified that some NSAIDs, including diclofenac, inhibit autophagic flux in hepatocytes. Further detailed studies demonstrated that diclofenac induced a reactive oxygen species (ROS)-dependent increase in lysosomal pH, attenuated cathepsin activity and blocked autophagosome-lysosome fusion. The reactivation of lysosomal function by treatment with clioquinol or transfection with the transcription factor EB restored lysosomal pH and thus autophagic flux. The production of mitochondrial ROS is critical for this process since scavenging ROS reversed lysosomal dysfunction and activated autophagic flux. The compromised lysosomal activity induced by diclofenac also inhibited the fusion with and degradation of mitochondria by mitophagy. Diclofenac-induced cell death and hepatotoxicity were effectively protected by rapamycin. Thus, we demonstrated that diclofenac induces the intracellular ROS production and lysosomal dysfunction that lead to the suppression of autophagy. Impaired autophagy fails to maintain mitochondrial integrity and aggravates the cellular ROS burden, which leads to diclofenac-induced hepatotoxicity.
Sigma-Aldrich
Diclofenac sodium salt
Pricing and availability is not currently available.
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Pricing and availability is not currently available.