Merck
  • Learning in a simple biological system: a pilot study of classical conditioning of human macrophages in vitro.
Learning in a simple biological system: a pilot study of classical conditioning of human macrophages in vitro.
Behavioral and brain functions : BBF (2011-11-22)
Gustav Nilsonne, Alva Appelgren, John Axelsson, Mats Fredrikson, Mats Lekander
ABSTRACT
Recent advances in cell biology and gene regulation suggest mechanisms whereby associative learning could be performed by single cells. Therefore, we explored a model of classical conditioning in human macrophages in vitro. In macrophage cultures, bacterial lipopolysaccharide (LPS; unconditioned stimulus) was paired once with streptomycin (conditioned stimulus). Secretion of interleukin-6 (IL-6) was used as response measure. At evocation, conditioning was not observed. Levels of IL-6 were higher only in those cultures that had been exposed to LPS in the learning phase (p's < .05), regardless whether they received the conditioned stimulus or not at evocation.However, habituation was evident, with a 62% loss of the IL-6 response after three LPS presentations (p < .001). If further experiments confirm that simple learning can occur in immune cells, this may have bearings not only on immune regulation, but also on the brain response to molecular signals detected in the periphery. Importantly, whether capacities for simple learning in single cells extend beyond habituation, and how this would be demonstrated, remain open questions.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Iscove′s Modified Dulbecco′s Medium, liquid, sterile-filtered, With sodium bicarbonate, without L-glutamine, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Iscove′s Modified Dulbecco′s Medium, With L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
Iscove′s Modified Dulbecco′s Medium, With sodium bicarbonate and L-glutamine, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
StableCell IMDM, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
Iscove′s Modified Dulbecco′s Medium, liquid