Merck
  • RCC2 Promotes Esophageal Cancer Growth by Regulating Activity and Expression of the Sox2 Transcription Factor.
RCC2 Promotes Esophageal Cancer Growth by Regulating Activity and Expression of the Sox2 Transcription Factor.
Molecular cancer research : MCR (2020-08-18)
Ali Calderon-Aparicio, Hiroyuki Yamamoto, Humberto De Vitto, Tianshun Zhang, Qiushi Wang, Ann M Bode, Zigang Dong
ABSTRACT
Regulator of chromosome condensation 2 (RCC2) is a protein located in the centrosome, which ensures that cell division proceeds properly. Previous reports show that RCC2 is overexpressed in some cancers and could play a key role in tumor development, but the mechanisms concerning how this occurs are not understood. Furthermore, no evidence exists regarding its role in esophageal cancer. We studied the relevance of RCC2 in esophageal cancer growth and its regulation on Sox2, an important transcription factor promoting esophageal cancer. RCC2 was overexpressed in esophageal tumors compared with normal tissue, and this overexpression was associated with tumorigenicity by increasing cell proliferation, anchorage-independent growth, and migration. These oncogenic effects were accompanied by overexpression of Sox2. RCC2 upregulated and stabilized Sox2 expression and its target genes by inhibiting ubiquitination-mediated proteasome degradation. Likewise, RCC2 increased the transcriptional activity and promoter binding of Sox2. In vivo studies indicated that RCC2 and Sox2 were overexpressed in esophageal tumors compared with normal tissue, and this upregulation occurs in the esophageal basal cell layer for both proteins. In conditional knockout mice, RCC2 deletion decreased the tumor nodule formation and progression in the esophagus compared with wild-type mice. Proliferating cell nuclear antigen expression, a cell proliferation marker, was also downregulated in RCC2 knockout mice. Overall, our data show for the first time that RCC2 is an important protein for the stabilization and transcriptional activation of Sox2 and further promotion of malignancy in esophageal cancer. IMPLICATIONS: This study shows that RCC2 controls Sox2 expression and transcriptional activity to mediate esophageal cancer formation.
Product Description
Supelco
EPA 8260A Surrogate Standards Mix, certified reference material, 2000 μg/mL each component in methanol
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)