Merck
  • The ABCG2 Q141K hyperuricemia and gout associated variant illuminates the physiology of human urate excretion.
The ABCG2 Q141K hyperuricemia and gout associated variant illuminates the physiology of human urate excretion.
Nature communications (2020-06-04)
Kazi Mirajul Hoque, Eryn E Dixon, Raychel M Lewis, Jordyn Allan, Gregory D Gamble, Amanda J Phipps-Green, Victoria L Halperin Kuhns, Anne M Horne, Lisa K Stamp, Tony R Merriman, Nicola Dalbeth, Owen M Woodward
ABSTRACT
The pathophysiological nature of the common ABCG2 gout and hyperuricemia associated variant Q141K (rs2231142) remains undefined. Here, we use a human interventional cohort study (ACTRN12615001302549) to understand the physiological role of ABCG2 and find that participants with the Q141K ABCG2 variant display elevated serum urate, unaltered FEUA, and significant evidence of reduced extra-renal urate excretion. We explore mechanisms by generating a mouse model of the orthologous Q140K Abcg2 variant and find male mice have significant hyperuricemia and metabolic alterations, but only subtle alterations of renal urate excretion and ABCG2 abundance. By contrast, these mice display a severe defect in ABCG2 abundance and function in the intestinal tract. These results suggest a tissue specific pathobiology of the Q141K variant, support an important role for ABCG2 in urate excretion in both the human kidney and intestinal tract, and provide insight into the importance of intestinal urate excretion for serum urate homeostasis.
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Mouse IGF-I ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6, clone C464.6, Upstate®, from mouse
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)