Merck
  • Tetrameric architecture of an active phenol-bound form of the AAA+ transcriptional regulator DmpR.
Tetrameric architecture of an active phenol-bound form of the AAA+ transcriptional regulator DmpR.
Nature communications (2020-06-03)
Kwang-Hyun Park, Sungchul Kim, Su-Jin Lee, Jee-Eun Cho, Vinod Vikas Patil, Arti Baban Dumbrepatil, Hyung-Nam Song, Woo-Chan Ahn, Chirlmin Joo, Seung-Goo Lee, Victoria Shingler, Eui-Jeon Woo
ABSTRACT
The Pseudomonas putida phenol-responsive regulator DmpR is a bacterial enhancer binding protein (bEBP) from the AAA+ ATPase family. Even though it was discovered more than two decades ago and has been widely used for aromatic hydrocarbon sensing, the activation mechanism of DmpR has remained elusive. Here, we show that phenol-bound DmpR forms a tetramer composed of two head-to-head dimers in a head-to-tail arrangement. The DmpR-phenol complex exhibits altered conformations within the C-termini of the sensory domains and shows an asymmetric orientation and angle in its coiled-coil linkers. The structural changes within the phenol binding sites and the downstream ATPase domains suggest that the effector binding signal is propagated through the coiled-coil helixes. The tetrameric DmpR-phenol complex interacts with the σ54 subunit of RNA polymerase in presence of an ATP analogue, indicating that DmpR-like bEBPs tetramers utilize a mechanistic mode distinct from that of hexameric AAA+ ATPases to activate σ54-dependent transcription.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Dextrose monohydrate, 97.5-102.0% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Cy®5 Maleimide Mono-Reactive Dye, Cytiva PA15131, pack of 1 mg