Merck
  • Synergistic lipid compositions for albumin receptor mediated delivery of mRNA to the liver.
Synergistic lipid compositions for albumin receptor mediated delivery of mRNA to the liver.
Nature communications (2020-05-18)
Lei Miao, Jiaqi Lin, Yuxuan Huang, Linxian Li, Derfogail Delcassian, Yifan Ge, Yunhua Shi, Daniel G Anderson
ABSTRACT
Lipid-like nanoparticles (LNPs) have potential as non-viral delivery systems for mRNA therapies. However, repeated administrations of LNPs may lead to accumulation of delivery materials and associated toxicity. To address this challenge, we have developed biodegradable lipids which improve LNPs clearance and reduce toxicity. We modify the backbone structure of Dlin-MC3-DMA by introducing alkyne and ester groups into the lipid tails. We evaluate the performance of these lipids when co-formulated with other amine containing lipid-like materials. We demonstrate that these formulations synergistically facilitate robust mRNA delivery with improved tolerability after single and repeated administrations. We further identify albumin-associated macropinocytosis and endocytosis as an ApoE-independent LNP cellular uptake pathway in the liver. Separately, the inclusion of alkyne lipids significantly increases membrane fusion to enhance mRNA release, leading to synergistic improvement of mRNA delivery. We believe that the rational design of LNPs with multiple amine-lipids increases the material space for mRNA delivery.
Sigma-Aldrich
MISSION® esiRNA, targeting human CLTC
Sigma-Aldrich
MISSION® esiRNA, targeting human LDLR
Sigma-Aldrich
MISSION® esiRNA, targeting human CAV1