Merck
  • DONSON and FANCM associate with different replisomes distinguished by replication timing and chromatin domain.
DONSON and FANCM associate with different replisomes distinguished by replication timing and chromatin domain.
Nature communications (2020-08-10)
Jing Zhang, Marina A Bellani, Ryan C James, Durga Pokharel, Yongqing Zhang, John J Reynolds, Gavin S McNee, Andrew P Jackson, Grant S Stewart, Michael M Seidman
ABSTRACT
Duplication of mammalian genomes requires replisomes to overcome numerous impediments during passage through open (eu) and condensed (hetero) chromatin. Typically, studies of replication stress characterize mixed populations of challenged and unchallenged replication forks, averaged across S phase, and model a single species of "stressed" replisome. Here, in cells containing potent obstacles to replication, we find two different lesion proximal replisomes. One is bound by the DONSON protein and is more frequent in early S phase, in regions marked by euchromatin. The other interacts with the FANCM DNA translocase, is more prominent in late S phase, and favors heterochromatin. The two forms can also be detected in unstressed cells. ChIP-seq of DNA associated with DONSON or FANCM confirms the bias of the former towards regions that replicate early and the skew of the latter towards regions that replicate late.
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Anti-DONSON antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FANCM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human DONSON
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)