Merck
  • Granzyme B Attenuates Bacterial Virulence by Targeting Secreted Factors.
Granzyme B Attenuates Bacterial Virulence by Targeting Secreted Factors.
iScience (2020-03-11)
Diego López León, Patricia Matthey, Isabelle Fellay, Marianne Blanchard, Denis Martinvalet, Pierre-Yves Mantel, Luis Filgueira, Michael Walch
ABSTRACT
Pathogenic bacteria secrete virulence factors that interact with the human host to establish infections. The human immune system evolved multiple mechanisms to fight bacterial invaders, including immune proteases that were demonstrated to contribute crucially to antibacterial defense. Here we show that granzyme B degrades multiple secreted virulence mediators from Listeria monocytogenes, Salmonella typhimurium, and Mycobacteria tuberculosis. Pathogenic bacteria, when infected in the presence of granzyme B or granzyme-secreting killer cells, fail to grow in human macrophages and epithelial cells owing to their crippled virulence. A granzyme B-uncleavable mutant form of the major Listeria virulence factor, listeriolysin O, rescued the virulence defect in response to granzyme treatment. Hence, we link the degradation of a single factor with the observed decrease in virulent bacteria growth. Overall, we reveal here an innate immune barrier function of granzyme B by disrupting bacterial virulence to facilitate bacteria clearance by bystander immune and non-immune cells.
Sigma-Aldrich
3,4-Dichloroisocoumarin, serine protease inhibitor
Pricing and availability is not currently available.