Merck
  • Systematic Analysis of Targets of Pumilio-Mediated mRNA Decay Reveals that PUM1 Repression by DNA Damage Activates Translesion Synthesis.
Systematic Analysis of Targets of Pumilio-Mediated mRNA Decay Reveals that PUM1 Repression by DNA Damage Activates Translesion Synthesis.
Cell reports (2020-05-07)
Toshimichi Yamada, Naoto Imamachi, Katsutoshi Imamura, Kenzui Taniue, Takeshi Kawamura, Yutaka Suzuki, Masami Nagahama, Nobuyoshi Akimitsu
ABSTRACT
RNA-binding proteins (RBPs) play a pivotal role in gene expression by modulating the stability of transcripts. However, the identification of degradation target mRNAs of RBPs remains difficult. By the combined analysis of transcriptome-wide mRNA stabilities and the binding of mRNAs to human Pumilio 1 (PUM1), we identify 48 mRNAs that both bind to PUM1 and exhibit PUM1-dependent degradation. Analysis of changes in the abundance of PUM1 and its degradation target mRNAs in RNA-seq data indicate that DNA-damaging agents negatively regulate PUM1-mediated mRNA decay. Cells exposed to cisplatin have reduced PUM1 abundance and increased PCNA and UBE2A mRNAs encoding proteins involved in DNA damage tolerance by translesion synthesis (TLS). Cells overexpressing PUM1 exhibit impaired DNA synthesis and TLS and increased sensitivity to the cytotoxic effect of cisplatin. Thus, our method identifies target mRNAs of PUM1-mediated decay and reveals that cells respond to DNA damage by inhibiting PUM1-mediated mRNA decay to activate TLS.
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-PUM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse