Merck
  • Chronic mTOR activation induces a degradative smooth muscle cell phenotype.
Chronic mTOR activation induces a degradative smooth muscle cell phenotype.
The Journal of clinical investigation (2020-02-11)
Guangxin Li, Mo Wang, Alexander W Caulk, Nicholas A Cilfone, Sharvari Gujja, Lingfeng Qin, Pei-Yu Chen, Zehua Chen, Sameh Yousef, Yang Jiao, Changshun He, Bo Jiang, Arina Korneva, Matthew R Bersi, Guilin Wang, Xinran Liu, Sameet Mehta, Arnar Geirsson, Jeffrey R Gulcher, Thomas W Chittenden, Michael Simons, Jay D Humphrey, George Tellides
ABSTRACT
Smooth muscle cell (SMC) proliferation has been thought to limit the progression of thoracic aortic aneurysm and dissection (TAAD) because loss of medial cells associates with advanced disease. We investigated effects of SMC proliferation in the aortic media by conditional disruption of Tsc1, which hyperactivates mTOR complex 1. Consequent SMC hyperplasia led to progressive medial degeneration and TAAD. In addition to diminished contractile and synthetic functions, fate-mapped SMCs displayed increased proteolysis, endocytosis, phagocytosis, and lysosomal clearance of extracellular matrix and apoptotic cells. SMCs acquired a limited repertoire of macrophage markers and functions via biogenesis of degradative organelles through an mTOR/β-catenin/MITF-dependent pathway, but were distinguishable from conventional macrophages by an absence of hematopoietic lineage markers and certain immune effectors even in the context of hyperlipidemia. Similar mTOR activation and induction of a degradative SMC phenotype in a model of mild TAAD due to Fbn1 mutation greatly worsened disease with near-uniform lethality. The finding of increased lysosomal markers in medial SMCs from clinical TAAD specimens with hyperplasia and matrix degradation further supports the concept that proliferation of degradative SMCs within the media causes aortic disease, thus identifying mTOR-dependent phenotypic modulation as a therapeutic target for combating TAAD.
Sigma-Aldrich
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Macrophages/Monocytes Antibody, clone MOMA-2, clone MOMA-2, Chemicon®, from rat
Sigma-Aldrich
Anti-MiTF Antibody, clone 3D1.2, clone 3D1.2, from mouse