Merck
  • Analogs of methyl-piperidinopyrazole (MPP): antiestrogens with estrogen receptor alpha selective activity.
Analogs of methyl-piperidinopyrazole (MPP): antiestrogens with estrogen receptor alpha selective activity.
Bioorganic & medicinal chemistry letters (2008-11-19)
Hai-Bing Zhou, Kathryn E Carlson, Fabio Stossi, Benita S Katzenellenbogen, John A Katzenellenbogen
ABSTRACT
Methyl-piperidino-pyrazole (MPP), an estrogen receptor alpha (ERalpha)-selective antagonist we developed, has a basic side chain (BSC) attached to an ERalpha-selective agonist ligand, methyl-pyrazole-triol (MPT) through an ether linkage. To remove the possibility that metabolic cleavage of the BSC in MPP would regenerate MPT, we have replaced the N-piperidinylethoxy moiety with an N-piperidinylpropyl group, giving MPrP. This new analog retains the high ERalpha-selective binding affinity and antagonist potency of MPP.
Sigma-Aldrich
MPP+ iodide, ≥98% (HPLC), powder
Sigma-Aldrich
MPP dihydrochloride hydrate, ≥97% (HPLC), white, powder