Merck
  • A Single Amino Acid Substitution in STKc_GSK3 Kinase Conferring Semispherical Grains and Its Implications for the Origin of Triticumsphaerococcum.
A Single Amino Acid Substitution in STKc_GSK3 Kinase Conferring Semispherical Grains and Its Implications for the Origin of Triticumsphaerococcum.
The Plant cell (2020-02-16)
Xuejiao Cheng, Mingming Xin, Ruibin Xu, Zhaoyan Chen, Wenlong Cai, Lingling Chai, Huanwen Xu, Lin Jia, Zhiyu Feng, Zihao Wang, Huiru Peng, Yingyin Yao, Zhaorong Hu, Weilong Guo, Zhongfu Ni, Qixin Sun
ABSTRACT
Six subspecies of hexaploid wheat (Triticumaestivum) have been identified, but the origin of Indian dwarf wheat (Triticum sphaerococcum), the only subspecies with round grains, is currently unknown. Here, we isolated the grain-shape gene Tasg-D1 in Tsphaerococcum via positional cloning. Tasg-D1 encodes a Ser/Thr protein kinase glycogen synthase kinase3 (STKc_GSK3) that negatively regulates brassinosteroid signaling. Expression of TaSG-D1 and the mutant form Tasg-D1 in Arabidopsis (Arabidopsis thaliana) suggested that a single amino acid substitution in the Thr-283-Arg-284-Glu-285-Glu-286 domain of TaSG-D1 enhances protein stability in response to brassinosteroids, likely leading to formation of round grains in wheat. This gain-of-function mutation has pleiotropic effects on plant architecture and exhibits incomplete dominance. Haplotype analysis of 898 wheat accessions indicated that the origin of Tsphaerococcum in ancient India involved at least two independent mutations of TaSG-D1 Our results demonstrate that modest genetic changes in a single gene can induce dramatic phenotypic changes.
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
Epibrassinolide, ≥85%
Sigma-Aldrich
Brassinazole, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin (plant) antibody, Mouse monoclonal, clone 10-B3 (MAbGPa), purified from hybridoma cell culture