Merck
  • Dynamics of Asymmetric and Symmetric Divisions of Muscle Stem Cells In Vivo and on Artificial Niches.
Dynamics of Asymmetric and Symmetric Divisions of Muscle Stem Cells In Vivo and on Artificial Niches.
Cell reports (2020-03-12)
Brendan Evano, Sara Khalilian, Gilles Le Carrou, Geneviève Almouzni, Shahragim Tajbakhsh
ABSTRACT
Stem cells can be maintained through symmetric cell divisions (SCDs) and asymmetric cell divisions (ACDs). How and when these divisions occur in vivo in vertebrates is poorly understood. Here, we developed a clonogenic cell tracing method that demonstrates the asymmetric distribution of transcription factors along with old and new DNA in mouse muscle stem cells during skeletal muscle regeneration. Combining single-cell tracking and artificial niches ex vivo, we show how cells switch from ACDs to SCDs, suggesting that they are not engaged in an obligate mode of cell division. Further, we generated SNAP-tagged histone H3-reporter mice and find that, unlike fly germline stem cells, differential fate outcomes are associated with a symmetric distribution of the H3.1 and H3.3 histone variants in mouse muscle stem cells. This versatile and efficient H3-SNAP labeling system will allow an investigation of mechanisms underlying the maintenance of epigenomic identity and plasticity in a variety of tissues.
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
MCDB 201 Medium, With trace elements, L-glutamine and 30 mM HEPES, powder, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Anti-phospho-Histone H3 (Thr3) Antibody, clone JY325, rabbit monoclonal, clone JY325, from rabbit
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g