Merck
  • Noncanonical regulation of phosphatidylserine metabolism by a Sec14-like protein and a lipid kinase.
Noncanonical regulation of phosphatidylserine metabolism by a Sec14-like protein and a lipid kinase.
The Journal of cell biology (2020-04-19)
Yaxi Wang, Peihua Yuan, Aby Grabon, Ashutosh Tripathi, Dongju Lee, Martin Rodriguez, Max Lönnfors, Michal Eisenberg-Bord, Zehua Wang, Sin Man Lam, Maya Schuldiner, Vytas A Bankaitis
ABSTRACT
The yeast phosphatidylserine (PtdSer) decarboxylase Psd2 is proposed to engage in a membrane contact site (MCS) for PtdSer decarboxylation to phosphatidylethanolamine (PtdEtn). This proposed MCS harbors Psd2, the Sec14-like phosphatidylinositol transfer protein (PITP) Sfh4, the Stt4 phosphatidylinositol (PtdIns) 4-OH kinase, the Scs2 tether, and an uncharacterized protein. We report that, of these components, only Sfh4 and Stt4 regulate Psd2 activity in vivo. They do so via distinct mechanisms. Sfh4 operates via a mechanism for which its PtdIns-transfer activity is dispensable but requires an Sfh4-Psd2 physical interaction. The other requires Stt4-mediated production of PtdIns-4-phosphate (PtdIns4P), where Stt4 (along with the Sac1 PtdIns4P phosphatase and endoplasmic reticulum-plasma membrane tethers) indirectly modulate Psd2 activity via a PtdIns4P homeostatic mechanism that influences PtdSer accessibility to Psd2. These results identify an example in which the biological function of a Sec14-like PITP is cleanly uncoupled from its canonical in vitro PtdIns-transfer activity and challenge popular functional assumptions regarding lipid-transfer protein involvements in MCS function.
Sigma-Aldrich
1-Naphthalenesulfonic acid, >50%
Avanti
16:0-d31-18:1 PS, Avanti Research - A Croda Brand 860403C
Avanti
16:0-d31-18:1 PS, Avanti Research - A Croda Brand 860403P, powder
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Avanti
14:0 PE, Avanti Research - A Croda Brand
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L, 1 mg/mL (after reconstitution), Chemicon®