Merck
  • mTORC1 Regulates Mitochondrial Integrated Stress Response and Mitochondrial Myopathy Progression.
mTORC1 Regulates Mitochondrial Integrated Stress Response and Mitochondrial Myopathy Progression.
Cell metabolism (2017-08-03)
Nahid A Khan, Joni Nikkanen, Shuichi Yatsuga, Christopher Jackson, Liya Wang, Swagat Pradhan, Riikka Kivelä, Alberto Pessia, Vidya Velagapudi, Anu Suomalainen
ABSTRACT
Mitochondrial dysfunction elicits various stress responses in different model systems, but how these responses relate to each other and contribute to mitochondrial disease has remained unclear. Mitochondrial myopathy (MM) is the most common manifestation of adult-onset mitochondrial disease and shows a multifaceted tissue-specific stress response: (1) transcriptional response, including metabolic cytokines FGF21 and GDF15; (2) remodeling of one-carbon metabolism; and (3) mitochondrial unfolded protein response. We show that these processes are part of one integrated mitochondrial stress response (ISRmt), which is controlled by mTORC1 in muscle. mTORC1 inhibition by rapamycin downregulated all components of ISRmt, improved all MM hallmarks, and reversed the progression of even late-stage MM, without inducing mitochondrial biogenesis. Our evidence suggests that (1) chronic upregulation of anabolic pathways contributes to MM progression, (2) long-term induction of ISRmt is not protective for muscle, and (3) rapamycin treatment trials should be considered for adult-type MM with raised FGF21.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Anti-LONP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Actinonin