Merck
  • In vitro methods to prepare astrocyte and motoneuron cultures for the investigation of potential in vivo interactions.
In vitro methods to prepare astrocyte and motoneuron cultures for the investigation of potential in vivo interactions.
Nature protocols (2007-06-05)
Anna R Taylor, Mac B Robinson, Carol E Milligan
ABSTRACT
This protocol details methods to isolate and purify astrocytes and motoneurons (MNs) from the chick lumbar spinal cord. In addition, an approach to study the influences of astrocyte secreted factors on MNs is provided. Astrocytes are isolated between embryonic days 10 and 12 (E10-12), propagated in serum (2-3 h) and differentiated in chemically defined medium (3-4 h). When prepared according to this protocol, astrocyte cultures are more than 98% pure when assessed using the astrocyte-specific markers glial fibrillary acidic protein (GFAP) and S100beta. MNs are isolated between E5.5 and 6.0 (3-4 h) using a procedure that takes selective advantage of the large size of these cells. These cultures can be maintained using individual trophic factors, target-derived factors or astrocyte-derived factors, the preparation of which is also described (5-6 h). All or part of these techniques can be used to investigate a variety of processes that occur during nervous system development and disease or after injury.
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Poly-DL-ornithine hydrobromide, mol wt 15,000-30,000