Merck
  • CoPP-Induced-Induced HO-1 Overexpression Alleviates Photoreceptor Degeneration With Rapid Dynamics: A Therapeutic Molecular Against Retinopathy.
CoPP-Induced-Induced HO-1 Overexpression Alleviates Photoreceptor Degeneration With Rapid Dynamics: A Therapeutic Molecular Against Retinopathy.
Investigative ophthalmology & visual science (2019-12-12)
Ye Tao, Lun Cai, Dawei Zhou, Chunhui Wang, Zhao Ma, Xiaofei Dong, Guanghua Peng
ABSTRACT
Retinitis pigmentosa (RP) causes progressive photoreceptor degeneration in the retina. The N-methyl-N-nitrosourea (MNU)-administered mouse is used as a chemically induced RP model with rapid progression rate. This study was designed to study heme oxygenase-1 (HO-1) expression in the MNU-administered mice, and to explore the therapeutic effects of cobalt protoporphyrin (CoPP). The HO-1 expression in the retina of MNU-administered mice was analyzed. CoPP was injected intravenously into the MNU-administered mice. Subsequently, the CoPP-treated mice were subjected to functional and morphologic examinations. HO-1 was involved in the MNU-induced photoreceptor degeneration. CoPP treatment enhanced retinal HO-1 expression in the MNU-administered mice. Electroretinogram (ERG) examination and behavioral tests showed that CoPP treatment improved the retinal responsiveness of MNU-administered mice. Histologic analysis and optical coherence tomography (OCT) examination showed that retinal architecture of the CoPP-treated mice was more intact than that of the MNU+vehicle group. Cone photoreceptors in the MNU-administered mice were rescued efficiently by CoPP treatment. Furthermore, multielectrode array (MEA) recording showed that CoPP treatment mitigated the spontaneous firing response, enhanced the light-induced firing response, and preserved the basic configurations of visual signal pathway in the MNU-administered mice. Mechanism studies suggested that CoPP afforded these therapeutic effects by modulating the apoptosis cascades and alleviating the oxidative stress in degenerative retinas. CoPP alleviated photoreceptor degeneration and rectified the signaling abnormities in MNU-administered mice. CoPP may serve as a potential medication against degenerative retinopathy.
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Protoporphyrin IX cobalt chloride
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit