Merck
  • The VNTR 48 bp Polymorphism in the DRD4 Gene Is Associated with Higher Tobacco Smoking in Male Mexican Mestizo Smokers with and without COPD.
The VNTR 48 bp Polymorphism in the DRD4 Gene Is Associated with Higher Tobacco Smoking in Male Mexican Mestizo Smokers with and without COPD.
Diagnostics (Basel, Switzerland) (2020-01-08)
Gloria Pérez-Rubio, Salvador García-Carmona, Leonor García-Gómez, Andrea Hernández-Pérez, Alejandra Ramírez-Venegas, Luis Alberto López-Flores, Raúl Sansores, Ramcés Falfán-Valencia
ABSTRACT
Cigarette smoking is influenced by nicotine's effects on dopaminergic activity, which appear to be moderated by genetic variation, particularly a variable number tandem repeat (VNTR, 48 bp) polymorphism in the third exon of the dopamine receptor gene (DRD4). Smokers with the VNTR ≥7 repeats (long, L allele) report markedly increased participation in some smoking behaviors; hence, our aim was to evaluate the association of the L allele in Mexican Mestizo smokers with and without COPD. The DRD4 VNTR 48 bp was genotyped in 492 Mexican Mestizo smokers: 164 COPD patients (≥20 cigarettes per day, cpd), 164 heavy smokers without COPD (HS, ≥20 cpd) and 164 light smokers without COPD (LS, 1-10 cpd). In the dominant model analysis (SL + LL vs. SS), men in the COPD and HS groups showed a statistical difference compared to LS (p = 0.01, OR = 2.06, CI 95% 1.17-3.64 and p = 0.05, OR = 1.88, CI 95% 1.03-3.45, respectively). In addition, by clustering smokers >20 cpd (COPD + HS) and comparing with the LS group, we found an association with increased risk of higher tobacco smoking p = 0.01, OR = 1.99, CI 95% 1.18-3.34. In conclusion, the long allele (L) in the VNTR of the DRD4 gene is associated with the risk of presenting higher tobacco smoking in male Mexican Mestizo smokers.
Product Description
Roche
KAPA Taq EXtra PCR Kit
Pricing and availability is not currently available.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl sulfoxide, ≥99%, FG
Pricing and availability is not currently available.