Merck
Spectral Imaging for Microbubble Characterization.
Langmuir : the ACS journal of surfaces and colloids (2019-12-20)
Richard J Browning, Miles Aron, Anna Booth, Paul Rademeyer, Sarah Wing, Veerle Brans, Shamit Shrivastava, Dario Carugo, Eleanor Stride
ABSTRACT
Microbubbles stabilized by an outer lipid shell have been studied extensively for both diagnostic and therapeutic applications. The shell composition can significantly influence microbubble behavior, but performing quantitative measurements of shell properties is challenging. The aim of this study is to investigate the use of spectral imaging to characterize the surface properties of a range of microbubble formulations representing both commercial and research agents. A lipophilic dye, C-laurdan, whose fluorescence emission varies according to the properties of the local environment, was used to compare the degree and uniformity of the lipid order in the microbubble shell, and these measurements were compared with the acoustic response and stability of the different formulations. Spectral imaging was found to be suitable for performing rapid and hence relatively high throughput measurements of microbubble surface properties. Interestingly, despite significant differences in lipid molecule size and charge, all of the different formulations exhibited highly ordered lipid shells. Measurements of liposomes with the same composition and the debris generated by destroying lipid microbubbles with ultrasound (US) showed that these exhibited a lower and more varied lipid order than intact microbubbles. This suggests that the high lipid order of microbubbles is due primarily to compression of the shell as a result of surface tension and is only minimally affected by composition. This also explains the similarity in acoustic response observed between the formulations, because microbubble dynamics are determined by the diameter and shell viscoelastic properties that are themselves a function of the lipid order. Within each population, there was considerable variability in the lipid order and response between individual microbubbles, suggesting the need for improved manufacturing techniques. In addition, the difference in the lipid order between the shell and lipid debris may be important for therapeutic applications in which shedding of the shell material is exploited, for example, drug delivery.
Product Description
Avanti
16:0 PA, Avanti Research - A Croda Brand
Avanti
18:0 EPC (Cl Salt), Avanti Research - A Croda Brand 890703C
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PEG5000 PE, Avanti Research - A Croda Brand 880220P, powder
Avanti
18:0 EPC (Cl Salt), Avanti Research - A Croda Brand 890703P, powder