Merck
  • Extended synaptotagmins are Ca2+-dependent lipid transfer proteins at membrane contact sites.
Extended synaptotagmins are Ca2+-dependent lipid transfer proteins at membrane contact sites.
Proceedings of the National Academy of Sciences of the United States of America (2016-04-05)
Haijia Yu, Yinghui Liu, Daniel R Gulbranson, Alex Paine, Shailendra S Rathore, Jingshi Shen
ABSTRACT
Organelles are in constant communication with each other through exchange of proteins (mediated by trafficking vesicles) and lipids [mediated by both trafficking vesicles and lipid transfer proteins (LTPs)]. It has long been known that vesicle trafficking can be tightly regulated by the second messenger Ca(2+), allowing membrane protein transport to be adjusted according to physiological demands. However, it remains unclear whether LTP-mediated lipid transport can also be regulated by Ca(2+) In this work, we show that extended synaptotagmins (E-Syts), poorly understood membrane proteins at endoplasmic reticulum-plasma membrane contact sites, are Ca(2+)-dependent LTPs. Using both recombinant and endogenous mammalian proteins, we discovered that E-Syts transfer glycerophospholipids between membrane bilayers in the presence of Ca(2+) E-Syts use their lipid-accommodating synaptotagmin-like mitochondrial lipid binding protein (SMP) domains to transfer lipids. However, the SMP domains themselves cannot transport lipids unless the two membranes are tightly tethered by Ca(2+)-bound C2 domains. Strikingly, the Ca(2+)-regulated lipid transfer activity of E-Syts was fully recapitulated when the SMP domain was fused to the cytosolic domain of synaptotagmin-1, the Ca(2+)sensor in synaptic vesicle fusion, indicating that a common mechanism of membrane tethering governs the Ca(2+)regulation of lipid transfer and vesicle fusion. Finally, we showed that microsomal vesicles isolated from mammalian cells contained robust Ca(2+)-dependent lipid transfer activities, which were mediated by E-Syts. These findings established E-Syts as a novel class of LTPs and showed that LTP-mediated lipid trafficking, like vesicular transport, can be subject to tight Ca(2+)regulation.
Product Description
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133P, powder
Pricing and availability is not currently available.
Avanti
18:1-12:0 NBD PC, Avanti Research - A Croda Brand 810133C
Pricing and availability is not currently available.
Avanti
18:1 NBD PS, Avanti Research - A Croda Brand 810198C
Pricing and availability is not currently available.