Merck
  • A cooperative activation loop among SWI/SNF, gamma-H2AX and H3 acetylation for DNA double-strand break repair.
A cooperative activation loop among SWI/SNF, gamma-H2AX and H3 acetylation for DNA double-strand break repair.
The EMBO journal (2010-03-13)
Han-Sae Lee, Ji-Hye Park, So-Jung Kim, Su-Jung Kwon, Jongbum Kwon
ABSTRACT
Although recent studies highlight the importance of histone modifications and ATP-dependent chromatin remodelling in DNA double-strand break (DSB) repair, how these mechanisms cooperate has remained largely unexplored. Here, we show that the SWI/SNF chromatin remodelling complex, earlier known to facilitate the phosphorylation of histone H2AX at Ser-139 (S139ph) after DNA damage, binds to gamma-H2AX (the phosphorylated form of H2AX)-containing nucleosomes in S139ph-dependent manner. Unexpectedly, BRG1, the catalytic subunit of SWI/SNF, binds to gamma-H2AX nucleosomes by interacting with acetylated H3, not with S139ph itself, through its bromodomain. Blocking the BRG1 interaction with gamma-H2AX nucleosomes either by deletion or overexpression of the BRG1 bromodomain leads to defect of S139ph and DSB repair. H3 acetylation is required for the binding of BRG1 to gamma-H2AX nucleosomes. S139ph stimulates the H3 acetylation on gamma-H2AX nucleosomes, and the histone acetyltransferase Gcn5 is responsible for this novel crosstalk. The H3 acetylation on gamma-H2AX nucleosomes is induced by DNA damage. These results collectively suggest that SWI/SNF, gamma-H2AX and H3 acetylation cooperatively act in a feedback activation loop to facilitate DSB repair.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H2A (acidic patch) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys36) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys18) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys9) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys23) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®