Merck
  • Reactivation of Dormant Relay Pathways in Injured Spinal Cord by KCC2 Manipulations.
Reactivation of Dormant Relay Pathways in Injured Spinal Cord by KCC2 Manipulations.
Cell (2018-07-24)
Bo Chen, Yi Li, Bin Yu, Zicong Zhang, Benedikt Brommer, Philip Raymond Williams, Yuanyuan Liu, Shane Vincent Hegarty, Songlin Zhou, Junjie Zhu, Hong Guo, Yi Lu, Yiming Zhang, Xiaosong Gu, Zhigang He
ABSTRACT
Many human spinal cord injuries are anatomically incomplete but exhibit complete paralysis. It is unknown why spared axons fail to mediate functional recovery in these cases. To investigate this, we undertook a small-molecule screen in mice with staggered bilateral hemisections in which the lumbar spinal cord is deprived of all direct brain-derived innervation, but dormant relay circuits remain. We discovered that a KCC2 agonist restored stepping ability, which could be mimicked by selective expression of KCC2, or hyperpolarizing DREADDs, in the inhibitory interneurons between and around the staggered spinal lesions. Mechanistically, these treatments transformed this injury-induced dysfunctional spinal circuit to a functional state, facilitating the relay of brain-derived commands toward the lumbar spinal cord. Thus, our results identify spinal inhibitory interneurons as a roadblock limiting the integration of descending inputs into relay circuits after injury and suggest KCC2 agonists as promising treatments for promoting functional recovery after spinal cord injury.
Sigma-Aldrich
Quipazine maleate salt
Sigma-Aldrich
CP-101,606, ≥98% (HPLC)
Sigma-Aldrich
Clozapine N-oxide
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-K+/Cl- Cotransporter (KCC2) Antibody, Upstate®, from rabbit