Merck
Replicative senescence revisited.
The journals of gerontology. Series A, Biological sciences and medical sciences (2002-06-27)
Richard Marcotte, Eugenia Wang
ABSTRACT
Forty years after its discovery, replicative senescence remains a rich source of information about cell-cycle regulation and the progression from a normal to a transformed phenotype. Effectors of this growth-arrested state are being discovered at a great pace. This review discusses the latest findings on the players responsible for establishing replicative senescence, as well as the associated telomere shortening.
Sigma-Aldrich
Cellular Senescence Assay, Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.