Merck
  • H3 clipping activity of glutamate dehydrogenase is regulated by stefin B and chromatin structure.
H3 clipping activity of glutamate dehydrogenase is regulated by stefin B and chromatin structure.
The FEBS journal (2014-09-30)
Papita Mandal, Sakshi Chauhan, Raghuvir S Tomar
ABSTRACT
Glutamate dehydrogenase has been recently identified as a tissue-specific histone H3-specific clipping enzyme. We have previously shown that it cleaves free as well as chromatin-bound histone H3. However, the physiological significance of this enzyme is still not clear. The present study aimed to improve our understanding of its significance in vivo. Using biochemical and cell biological approaches, we show that glutamate dehydrogenase is primarily associated with euchromatin, and it re-localizes from the nuclear periphery to the nucleolus upon DNA damage. The cysteine protease inhibitor stefin B regulates the H3 clipping activity of the enzyme. Chromatin structure and certain histone modifications influence H3 clipping activity. Interestingly, we also observed that an in vivo truncated form of H3 lacks H3K56 acetylation, which is a code for the DNA damage response. Together, these results suggest that glutamate dehydrogenase is a euchromatin-associated enzyme, and its H3 clipping activity is regulated by chromatin structure, histone modifications and an in vivo inhibitor. In response to DNA damage, it re-localizes to the nuclei, and hence may be involved in regulation of gene expression in vivo.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-dimethyl-Histone H1.4 (diMe-Lys26) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Histone H4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-dimethyl-Histone H3 (diMe-Lys9) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-acetyl- & phospho-Histone H3 (Ac-Lys9, pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Fibrillarin antibody produced in rabbit, affinity isolated antibody