Merck
  • PIP2 increases the speed of response of synaptotagmin and steers its membrane-penetration activity toward the plasma membrane.
PIP2 increases the speed of response of synaptotagmin and steers its membrane-penetration activity toward the plasma membrane.
Nature structural & molecular biology (2004-01-14)
Jihong Bai, Ward C Tucker, Edwin R Chapman
ABSTRACT
Synaptotagmin-1 (syt), the putative Ca2+ sensor for exocytosis, is anchored to the membrane of secretory organelles. Its cytoplasmic domain is composed of two Ca2+-sensing modules, C2A and C2B. Syt binds phosphatidylinositol 4,5-bisphosphate (PIP2), a plasma membrane lipid with an essential role in exocytosis and endocytosis. We resolved two modes of PIP2 binding that are mediated by distinct surfaces on the C2B domain of syt. A novel Ca2+-independent mode of binding predisposes syt to penetrate PIP2-harboring target membranes in response to Ca2+ with submillisecond kinetics. Thus, PIP2 increases the speed of response of syt and steers its membrane-penetration activity toward the plasma membrane. We propose that syt-PIP2 interactions are involved in exocytosis by facilitating the close apposition of the vesicle and target membrane on rapid time scales in response to Ca2+.
Product Description
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
16:0-12 Doxyl PC, Avanti Research - A Croda Brand 810600P, powder
Avanti
Brain PI(4,5)P2, Avanti Research - A Croda Brand
Avanti
16:0-5 Doxyl PC, Avanti Research - A Croda Brand 810601C
Avanti
16:0-5 Doxyl PC, Avanti Research - A Croda Brand 810601P, powder
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 PS (DOPS), Avanti Research - A Croda Brand
Avanti
16:0-7 Doxyl PC, Avanti Research - A Croda Brand 810602P, powder